Assay Details
Target Gene Details
Entrez Gene ID: | 16183 |
Gene Name: | interleukin 2 |
Gene Aliases: |
Il-2 |
Location: |
Chr.3:37120713-37125954 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Il2 | NM_008366.3 | NP_032392.1 | ||
AF065914.1 | AAD25890.1 | |||
AF065915.1 | AAD25891.1 | |||
AF065916.1 | AAD25892.1 | |||
AY147902.1 | AAN38301.1 | |||
BC116845.1 | ||||
BC116873.1 | ||||
DQ836354.1 | ||||
K02292.1 | AAA39289.1 | |||
U41494.1 | AAB39206.1 | |||
U41504.1 | AAB39208.1 | |||
U41505.1 | AAB39209.1 | |||
U41506.1 | AAB39210.1 | |||
X01772.1 | CAA25909.1 | |||
X73040.1 |
More Information
Set Membership: |
Intragenic Non-exonic |