Assay Details
Target Gene Details
Entrez Gene ID: | 641365 |
Gene Name: | long intergenic non-protein coding RNA 616 |
Gene Aliases: |
- |
Location: |
Chr.4:138027422-138130685 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LINC00616 | NR_037866.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3564703 | Chr.4:138045004 - 138046097 on Build GRCh38 | Deletion |
![]() |
esv2728410 | Chr.4:138045016 - 138046095 on Build GRCh38 | Deletion |
![]() |
dgv2910n106 | Chr.4:138044946 - 138046065 on Build GRCh38 | Deletion |
![]() |
esv2667601 | Chr.4:138045057 - 138046064 on Build GRCh38 | Deletion |
![]() |
esv3602409 | Chr.4:138045035 - 138046056 on Build GRCh38 | Loss |
![]() |
esv1961455 | Chr.4:138044877 - 138046234 on Build GRCh38 | Deletion |
![]() |
dgv173n6 | Chr.4:138045058 - 138046064 on Build GRCh38 | Deletion |
![]() |
dgv152n73 | Chr.4:138045047 - 138046056 on Build GRCh38 | Deletion |
![]() |
nsv830080 | Chr.4:138031664 - 138223572 on Build GRCh38 | Gain |
![]() ![]() ![]() |
dgv127e180 | Chr.4:138044874 - 138046280 on Build GRCh38 | Deletion |
![]() |
nsv1033022 | Chr.4:137999933 - 138081308 on Build GRCh38 | Loss |
![]() |
nsv820431 | Chr.4:138044980 - 138046067 on Build GRCh38 | Deletion |
![]() |
esv2641901 | Chr.4:138044584 - 138046231 on Build GRCh38 | Deletion |
![]() |
nsv1149302 | Chr.4:138045053 - 138046054 on Build GRCh38 | Deletion |
![]() |
nsv513167 | Chr.4:138044504 - 138047172 on Build GRCh38 | Loss |
![]() |
nsv595579 | Chr.4:138045351 - 138047403 on Build GRCh38 | Loss |
![]() |
esv27865 | Chr.4:138045274 - 138046056 on Build GRCh38 | Gain+Loss |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)