Assay Details
Target Gene Details
Entrez Gene ID: | 4947 |
Gene Name: | ornithine decarboxylase antizyme 2 |
Gene Aliases: |
AZ2 |
Location: |
Chr.15:64687574-64703281 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
OAZ2 | NM_001301302.1 | NP_001288231.1 | ||
NM_002537.3 | NP_002528.1 | |||
AF057297.1 | AAD03265.1 | |||
AF242520.1 | ||||
AF242521.1 | AAF99601.1 | |||
BI597670.1 | ||||
DA210449.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv569702 | Chr.15:64702244 - 64706393 on Build GRCh38 | Loss |
![]() |
esv22620 | Chr.15:64702166 - 64702816 on Build GRCh38 | Loss |
![]() |
nsv569701 | Chr.15:64700971 - 64702760 on Build GRCh38 | Loss |
![]() |
nsv833038 | Chr.15:64522753 - 64724042 on Build GRCh38 | Loss |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)