Assay Details
Target Gene Details
Entrez Gene ID: | 8835 |
Gene Name: | suppressor of cytokine signaling 2 |
Gene Aliases: |
CIS2, Cish2, SOCS-2, SSI-2, SSI2, STATI2 |
Location: |
Chr.12:93569822-93626236 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SOCS2 | NM_001270467.1 | NP_001257396.1 | ||
NM_001270468.1 | NP_001257397.1 | |||
NM_001270469.1 | NP_001257398.1 | |||
NM_001270470.1 | 1 | 313 | NP_001257399.1 | |
NM_003877.4 | NP_003868.1 | |||
XM_017020147.1 | XP_016875636.1 | |||
XM_017020148.1 | XP_016875637.1 | |||
XM_017020149.1 | XP_016875638.1 | |||
XM_017020150.1 | XP_016875639.1 | |||
XM_017020151.1 | XP_016875640.1 | |||
XM_017020152.1 | XP_016875641.1 | |||
XM_017020154.1 | XP_016875643.1 | |||
XM_017020156.1 | 1 | 110 | XP_016875645.1 | |
AF037989.1 | AAC34745.1 | |||
AK290546.1 | ||||
AK297662.1 | ||||
AK309452.1 | ||||
AK313165.1 | ||||
AL522912.3 | ||||
DA490057.1 | ||||
DB054162.1 | ||||
DB481261.1 | 1 | 89 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv25615 | Chr.12:93570606 - 93572157 on Build GRCh38 | Loss | SOCS2 SOCS2-AS1 |
dgv2791n54 | Chr.12:93570630 - 93572036 on Build GRCh38 | Loss | SOCS2 SOCS2-AS1 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |