Assay Details
Target Gene Details
Entrez Gene ID: | 51084 |
Gene Name: | crystallin lambda 1 |
Gene Aliases: |
GDH, HEL30, lambda-CRY |
Location: |
Chr.13:20403667-20525873 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CRYL1 | NM_015974.2 | NP_057058.2 | ||
XM_005266415.3 | XP_005266472.1 | |||
XM_017020624.1 | XP_016876113.1 | |||
AF077049.1 | AAD27782.1 | |||
AF087898.1 | AAP97197.1 | |||
AF160216.1 | AAF67017.1 | |||
AK024041.1 | ||||
BC008562.2 | ||||
BC071810.1 | ||||
BC119660.1 | ||||
BC119661.1 | ||||
EU794604.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3892302 | Chr.13:20412913 - 20441076 on Build GRCh38 | Loss | CRYL1 MIR4499 |
esv34984 | Chr.13:20398561 - 20542402 on Build GRCh38 | Loss | CRYL1 MIR4499 |
dgv1612n100 | Chr.13:20222652 - 20530273 on Build GRCh38 | Loss | CRYL1 GJB6 MIR4499 |
nsv818947 | Chr.13:20401130 - 20521581 on Build GRCh38 | Loss | CRYL1 MIR4499 |
dgv3075n54 | Chr.13:20398414 - 20521581 on Build GRCh38 | Loss | CRYL1 MIR4499 |
esv27122 | Chr.13:20424213 - 20424798 on Build GRCh38 | Gain | CRYL1 |
dgv3072n54 | Chr.13:20216603 - 20521581 on Build GRCh38 | Loss | CRYL1 GJB6 MIR4499 |
nsv1046946 | Chr.13:19730301 - 20507454 on Build GRCh38 | Gain | CRYL1 PSPC1 GJA3 GJB6 ZMYM2 GJB2 LINC01072 MIR4499 ZMYM5 |
esv2759925 | Chr.13:20398583 - 20650339 on Build GRCh38 | Loss | CRYL1 IFT88 MIR4499 |
esv2751132 | Chr.13:20217943 - 20546665 on Build GRCh38 | Loss | CRYL1 GJB6 MIR4499 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |