Assay Details
Target Gene Details
Entrez Gene ID: | 51547 |
Gene Name: | sirtuin 7 |
Gene Aliases: |
SIR2L7 |
Location: |
Chr.17:81911939-81918182 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SIRT7 | NM_016538.2 | NP_057622.1 | ||
XM_006722284.2 | XP_006722347.1 | |||
XM_011523580.1 | XP_011521882.1 | |||
XM_011523581.2 | XP_011521883.1 | |||
AF233395.1 | AAF43431.1 | |||
AK002027.1 | BAA92044.1 | |||
AK094326.1 | ||||
AK131437.1 | ||||
AK131484.1 | 4 | 1680 | ||
AK290265.1 | ||||
AL137626.1 | CAB70848.2 | |||
BC017305.2 | AAH17305.1 | |||
BC101791.1 | AAI01792.1 | |||
BC101793.1 | AAI01794.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv29964 | Chr.17:81891311 - 81952513 on Build GRCh38 | Loss | NOTUM MYADML2 PCYT2 MAFG-AS1 ALYREF ANAPC11 PYCR1 NPB SIRT7 MAFG |
nsv833571 | Chr.17:81913391 - 81979943 on Build GRCh38 | Loss | NOTUM MYADML2 MAFG-AS1 ASPSCR1 PYCR1 SIRT7 MAFG |
nsv952392 | Chr.17:81885825 - 82065724 on Build GRCh38 | Deletion | GPS1 DUS1L NOTUM MYADML2 PCYT2 LRRC45 RAC3 STRA13 DCXR PYCR1 NPB RFNG MAFG-AS1 ASPSCR1 ALYREF ANAPC11 SIRT7 MAFG |
nsv428351 | Chr.17:81152631 - 83064073 on Build GRCh38 | Gain | LOC101929552 OGFOD3 FN3KRP LRRC45 FAAP100 DCXR CSNK1D PYCR1 PDE6G ARHGDIA RFNG MIR3186 UTS2R SNORD134 MIR6786 HEXDC NARF TEPSIN PPP1R27 LOC100130370 SIRT7 MIR6787 ARL16 TSPAN10 CEP131 MIR4525 PCYT2 OXLD1 P4HB LOC105371925 STRA13 CCDC137 MCRIP1 NPB FOXK2 MIR4740 WDR45B SLC25A10 LOC105371944 MAFG-AS1 ALYREF ANAPC11 TMEM105 LINC00482 MAFG CD7 GPS1 DUS1L RAB40B MYADML2 FSCN2 RAC3 SLC16A3 AATK HGS AATK-AS1 TEX19 ZNF750 LOC105376790 SECTM1 GCGR FN3K FASN NOTUM C17orf89 ACTG1 MRPL12 LOC105376791 LOC101929511 NPLOC4 CCDC57 BAHCC1 C17orf62 SLC38A10 TBCD B3GNTL1 ASPSCR1 |
nsv470620 | Chr.17:81516973 - 82064485 on Build GRCh38 | Loss | GPS1 DUS1L MYADML2 FSCN2 LRRC45 FAAP100 RAC3 DCXR PYCR1 HGS PDE6G ARHGDIA RFNG MIR6786 PPP1R27 GCGR SIRT7 ARL16 NOTUM TSPAN10 PCYT2 OXLD1 MRPL12 P4HB STRA13 CCDC137 MCRIP1 NPB NPLOC4 SLC25A10 MAFG-AS1 ASPSCR1 ALYREF ANAPC11 MAFG |
esv33468 | Chr.17:81891535 - 81927756 on Build GRCh38 | Loss | PCYT2 ALYREF ANAPC11 NPB SIRT7 MAFG |
nsv833569 | Chr.17:81838937 - 82011412 on Build GRCh38 | Loss | NOTUM MYADML2 PCYT2 P4HB PYCR1 NPB ARHGDIA MAFG-AS1 ASPSCR1 ALYREF ANAPC11 SIRT7 MAFG |
esv21913 | Chr.17:81828123 - 83103548 on Build GRCh38 | Gain+Loss | LOC101929552 OGFOD3 FN3KRP LRRC45 DCXR CSNK1D PYCR1 ARHGDIA RFNG UTS2R SNORD134 HEXDC NARF PPP1R27 SIRT7 MIR6787 MIR4525 PCYT2 P4HB STRA13 MCRIP1 NPB FOXK2 WDR45B LOC105371944 MAFG-AS1 ALYREF ANAPC11 MAFG CD7 GPS1 DUS1L RAB40B MYADML2 RAC3 SLC16A3 TEX19 METRNL ZNF750 LOC105376790 SECTM1 FN3K FASN NOTUM LOC105376791 LOC101929511 CCDC57 C17orf62 TBCD B3GNTL1 ASPSCR1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |