Assay Details
Target Gene Details
Entrez Gene ID: | 6358 |
Gene Name: | C-C motif chemokine ligand 14 |
Gene Aliases: |
CC-1, CC-3, CKB1, HCC-1, HCC-1(1-74), HCC-1/HCC-3, HCC-3, MCIF, NCC-2, NCC2, SCYA14, SCYL2, SY14 |
Location: |
Chr.17:35983656-35986728 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CCL14 | NM_032962.4 | NP_116738.1 | ||
NM_032963.3 | NP_116739.1 | |||
AA612866.1 | ||||
AK122606.1 | ||||
BC038289.1 | AAH38289.1 | |||
BC045165.1 | AAH45165.1 | |||
DA821276.1 | ||||
Z49270.1 | CAA89264.1 |
Target Gene Details
Entrez Gene ID: | 348249 |
Gene Name: | CCL15-CCL14 readthrough (NMD candidate) |
Gene Aliases: |
- |
Location: |
Chr.17:35983656-36002038 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CCL15-CCL14 | NR_027921.3 | |||
NR_027922.3 | ||||
Z70292.1 | ||||
Z70293.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1059795 | Chr.17:35828724 - 36157706 on Build GRCh38 | Gain | LYZL6 CCL23 CCL5 CCL15-CCL14 CCL3 LOC101927339 CCL18 TAF15 RDM1 CCL15 LRRC37A8P HEATR9 CCL14 CCL16 CCL4 |
nsv833427 | Chr.17:35958281 - 36115227 on Build GRCh38 | Loss | CCL23 CCL15-CCL14 CCL3 CCL14 LOC101927339 CCL18 CCL16 CCL15 CCL4 |
nsv833424 | Chr.17:35911883 - 36092128 on Build GRCh38 | Gain | LYZL6 CCL23 CCL15-CCL14 CCL3 CCL14 LOC101927339 CCL18 CCL16 RDM1 CCL15 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |