Assay Details
Target Gene Details
Entrez Gene ID: | 441161 |
Gene Name: | oocyte expressed protein |
Gene Aliases: |
C6orf156, FLOPED, HOEP19, KHDC2 |
Location: |
Chr.6:73368553-73369888 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
OOEP | NM_001080507.2 | NP_001073976.1 | ||
XM_017010890.1 | XP_016866379.1 | |||
BX093821.1 | ||||
EU290647.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv830693 | Chr.6:73339619 - 73506357 on Build GRCh38 | Loss | OOEP OOEP-AS1 DDX43 KHDC3L DPPA5 MTO1 MB21D1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |