Assay Details
Target Gene Details
Entrez Gene ID: | 105377934 |
Gene Name: | putative uncharacterized protein C6orf183 |
Gene Aliases: |
- |
Location: |
Chr.6:109165792-109203801 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC105377934 | XM_017011615.1 | XP_016867104.1 | ||
XM_017011616.1 | XP_016867105.1 | |||
XM_017011617.1 | XP_016867106.1 | |||
XM_017011618.1 | XP_016867107.1 | |||
XM_017011619.1 | XP_016867108.1 |