Assay Details
Target Gene Details
Entrez Gene ID: | 2259 |
Gene Name: | fibroblast growth factor 14 |
Gene Aliases: |
FGF-14, FHF-4, FHF4, SCA27 |
Location: |
Chr.13:101720855-102402428 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FGF14 | NM_001321937.1 | NP_001308866.1 | ||
NM_001321938.1 | NP_001308867.1 | |||
NM_001321939.1 | NP_001308868.1 | |||
NM_001321940.1 | NP_001308869.1 | |||
NM_001321941.1 | NP_001308870.1 | |||
NM_001321942.1 | NP_001308871.1 | |||
NM_001321943.1 | NP_001308872.1 | |||
NM_001321944.1 | NP_001308873.1 | |||
NM_001321945.1 | NP_001308874.1 | |||
NM_001321946.1 | NP_001308875.1 | |||
NM_001321947.1 | NP_001308876.1 | |||
NM_001321948.1 | NP_001308877.1 | |||
NM_001321949.1 | NP_001308878.1 | |||
NM_175929.2 | NP_787125.1 | |||
AY188178.1 | AAO31806.1 | |||
BC100920.1 | AAI00921.1 | |||
BC100921.1 | AAI00922.1 | |||
BC100922.2 | AAI00923.1 |
Target Gene Details
Entrez Gene ID: | 283481 |
Gene Name: | FGF14 antisense RNA 2 |
Gene Aliases: |
- |
Location: |
Chr.13:102394578-102395705 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FGF14-AS2 | NR_036487.1 | 1 | 350 | |
BX096285.1 | 1 | 350 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic non-DGV Variation |