Assay Details
Target Gene Details
Entrez Gene ID: | 149773 |
Gene Name: | APCDD1L antisense RNA 1 (head to head) |
Gene Aliases: |
- |
Location: |
Chr.20:58515379-58619892 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
APCDD1L-AS1 | NR_034147.1 | |||
AK054637.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1063462 | Chr.20:58511618 - 58790600 on Build GRCh38 | Gain | NPEPL1 STX16 APCDD1L LOC79160 LOC107985410 APCDD1L-AS1 STX16-NPEPL1 LOC105372695 |
nsv834019 | Chr.20:58397598 - 58607087 on Build GRCh38 | Loss | APCDD1L VAPB APCDD1L-AS1 |