Assay Details
Target Gene Details
Entrez Gene ID: | 23315 |
Gene Name: | solute carrier family 9 member A8 |
Gene Aliases: |
NHE-8, NHE8 |
Location: |
Chr.20:49812713-49892242 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3646017 | Chr.20:49610386 - 49857106 on Build GRCh38 | Gain | B4GALT5 SLC9A8 |
esv3646034 | Chr.20:49788665 - 49826482 on Build GRCh38 | Gain | SLC9A8 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |