Assay Details
Target Gene Details
Entrez Gene ID: | 191 |
Gene Name: | adenosylhomocysteinase |
Gene Aliases: |
SAHH, adoHcyase |
Location: |
Chr.20:34235012-34311976 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AHCY | XM_011528657.2 | XP_011526959.2 |
Target Gene Details
Entrez Gene ID: | 434 |
Gene Name: | agouti signaling protein |
Gene Aliases: |
AGSW, AGTI, AGTIL, ASP, SHEP9 |
Location: |
Chr.20:34186493-34269344 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ASIP | NM_001672.2 | NP_001663.2 | ||
XM_005260412.3 | XP_005260469.1 | |||
XM_011528820.2 | XP_011527122.1 | |||
XM_011528821.1 | XP_011527123.1 | |||
BC104238.1 | AAI04239.1 | |||
BC104239.2 | AAI04240.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1059756 | Chr.20:34143188 - 34368334 on Build GRCh38 | Gain | ASIP LOC107985381 AHCY ITCH |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |