Assay Details
Target Gene Details
Entrez Gene ID: | 84174 |
Gene Name: | Src like adaptor 2 |
Gene Aliases: |
C20orf156, MARS, SLAP-2, SLAP2 |
Location: |
Chr.20:36612519-36646216 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLA2 | NM_032214.3 | NP_115590.1 | ||
NM_175077.2 | NP_778252.1 | |||
XM_017028098.1 | XP_016883587.1 | |||
AF290985.1 | AAL38197.1 | |||
AF290986.1 | AAL38198.1 | |||
AF326353.1 | AAL29204.1 | |||
AK025645.1 | BAB15201.1 | |||
AK291513.1 | ||||
BC042041.1 | AAH42041.1 | |||
BQ052468.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833967 | Chr.20:36477427 - 36658869 on Build GRCh38 | Loss | SLA2 DLGAP4-AS1 TGIF2-C20orf24 NDRG3 MYL9 TGIF2 DLGAP4 C20orf24 |
nsv1057192 | Chr.20:36514051 - 36839310 on Build GRCh38 | Loss | SLA2 DLGAP4-AS1 SOGA1 TGIF2-C20orf24 NDRG3 MYL9 TGIF2 DLGAP4 C20orf24 DSN1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |