Assay Details
Target Gene Details
Entrez Gene ID: | 6714 |
Gene Name: | SRC proto-oncogene, non-receptor tyrosine kinase |
Gene Aliases: |
ASV, SRC1, THC6, c-SRC, p60-Src |
Location: |
Chr.20:37344685-37405432 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SRC | NM_005417.4 | NP_005408.1 | ||
NM_198291.2 | NP_938033.1 | |||
XM_011529013.2 | XP_011527315.1 | |||
XM_017028024.1 | XP_016883513.1 | |||
XM_017028025.1 | XP_016883514.1 | |||
XM_017028026.1 | XP_016883515.1 | |||
AF272982.1 | ||||
HY016645.1 | ||||
Z18364.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv7565n54 | Chr.20:37326534 - 37439471 on Build GRCh38 | Loss | SRC |
nsv523618 | Chr.20:37340671 - 37466486 on Build GRCh38 | Loss | SRC RPL7AP14 |
nsv954031 | Chr.20:37344798 - 37366197 on Build GRCh38 | Deletion | SRC |
nsv470551 | Chr.20:37314315 - 37448174 on Build GRCh38 | Gain | MANBAL SRC RPL7AP14 |
nsv524059 | Chr.20:37248607 - 37466486 on Build GRCh38 | Loss | MANBAL SRC GHRH RPL7AP14 |
nsv1055206 | Chr.20:37267421 - 37621089 on Build GRCh38 | Loss | LINC00489 MANBAL BLCAP SRC NNAT RPL7AP14 |
nsv509767 | Chr.20:37305577 - 37477600 on Build GRCh38 | Insertion | MANBAL SRC RPL7AP14 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |