Assay Details
Target Gene Details
Entrez Gene ID: | 23054 |
Gene Name: | nuclear receptor coactivator 6 |
Gene Aliases: |
AIB3, ASC2, NRC, PRIP, RAP250, TRBP |
Location: |
Chr.20:34714774-34825649 on Build GRCh38 |
Assay Gene Location: | Within Intron 19 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1066181 | Chr.20:34682373 - 34725674 on Build GRCh38 | Gain | NCOA6 TP53INP2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |