Assay Details
Target Gene Details
Entrez Gene ID: | 2686 |
Gene Name: | gamma-glutamyltransferase 7 |
Gene Aliases: |
D20S101, GGT4, GGTL3, GGTL5, dJ18C9.2 |
Location: |
Chr.20:34844720-34872858 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GGT7 | NM_178026.2 | NP_821158.2 | ||
XM_011528782.2 | XP_011527084.1 | |||
XM_011528783.2 | XP_011527085.1 | |||
XM_017027809.1 | XP_016883298.1 | |||
BC101370.1 | ||||
BC101371.1 | ||||
BC101372.1 | ||||
BC141843.1 | ||||
DA117929.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1055447 | Chr.20:34737641 - 35020344 on Build GRCh38 | Loss | ACSS2 NCOA6 MIR499A HMGB3P1 TRPC4AP MIR499B MYH7B GSS GGT7 |
esv3567955 | Chr.20:34838184 - 34853908 on Build GRCh38 | Loss | GGT7 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |