Assay Details
Target Gene Details
Entrez Gene ID: | 10047 |
Gene Name: | cystatin 8 |
Gene Aliases: |
CRES, CTES5 |
Location: |
Chr.20:23491077-23507124 on Build GRCh38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CST8 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv585723 | Chr.20:23446011 - 23588763 on Build GRCh38 | Gain | CST13P CST9L CST11 CST8 |
esv3645525 | Chr.20:23425607 - 23597079 on Build GRCh38 | Gain | CST13P CST9L CST11 CSTL1 CST8 |
nsv1065390 | Chr.20:23432253 - 23588601 on Build GRCh38 | Loss | CST13P CST9L CST11 CSTL1 CST8 |
nsv470544 | Chr.20:23444001 - 23588763 on Build GRCh38 | Gain | CST13P CST9L CST11 CSTL1 CST8 |
nsv1063655 | Chr.20:23416012 - 23509093 on Build GRCh38 | Gain | NAPB CST11 CSTL1 CST8 |