Assay Details
Target Gene Details
Entrez Gene ID: | 57446 |
Gene Name: | NDRG family member 3 |
Gene Aliases: |
- |
Location: |
Chr.20:36651766-36746138 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NDRG3 | NM_022477.3 | NP_071922.2 | ||
NM_032013.3 | NP_114402.1 | |||
NR_038370.1 | ||||
XM_006723837.3 | XP_006723900.1 | |||
XM_006723838.3 | XP_006723901.1 | |||
XM_011528925.2 | XP_011527227.1 | |||
XM_011528926.2 | XP_011527228.1 | |||
XM_011528927.2 | XP_011527229.1 | |||
XM_011528928.2 | XP_011527230.1 | |||
XM_017027978.1 | XP_016883467.1 | |||
XM_017027979.1 | XP_016883468.1 | |||
XM_017027980.1 | XP_016883469.1 | |||
AB044943.1 | BAB20067.1 | |||
AF308609.1 | AAL08807.1 | |||
AK023618.1 | BAB14620.1 | |||
AK027665.1 | BAB55277.1 | |||
AK295420.1 | ||||
AK295466.1 | ||||
BC136489.1 | ||||
BC144169.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833967 | Chr.20:36477427 - 36658869 on Build GRCh38 | Loss | TGIF2 MYL9 DLGAP4-AS1 DLGAP4 NDRG3 C20orf24 SLA2 TGIF2-C20orf24 |
nsv1057192 | Chr.20:36514051 - 36839310 on Build GRCh38 | Loss | SOGA1 DSN1 TGIF2 MYL9 DLGAP4-AS1 DLGAP4 NDRG3 C20orf24 SLA2 TGIF2-C20orf24 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |