Assay Details
Target Gene Details
Entrez Gene ID: | 4784 |
Gene Name: | nuclear factor I X |
Gene Aliases: |
MRSHSS, NF1A, SOTOS2 |
Location: |
Chr.19:12995512-13098796 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NFIX | NM_002501.3 | NP_002492.2 | ||
XM_005259918.4 | XP_005259975.1 | |||
XM_017026837.1 | XP_016882326.1 | |||
BF115883.1 | ||||
BT019732.1 | AAV38537.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv828444 | Chr.19:13012681 - 13027353 on Build GRCh38 | Loss | NFIX |
nsv1055900 | Chr.19:12979147 - 13034197 on Build GRCh38 | Gain | NFIX |
nsv953977 | Chr.19:12666887 - 13095486 on Build GRCh38 | Deletion | FARSA DNASE2 ASNA1 SNORD135 DAND5 MIR6794 SNORD41 FBXW9 JUNB GADD45GIP1 NFIX PRDX2 C19orf43 CALR WDR83 RAD23A DHPS BEST2 MIR6515 RTBDN WDR83OS LOC105372280 KLF1 RNASEH2A SYCE2 HOOK2 MIR5695 MAST1 TNPO2 GCDH MIR5684 |
nsv2422 | Chr.19:13000362 - 13032711 on Build GRCh38 | Insertion | NFIX |
nsv833754 | Chr.19:12876208 - 13028186 on Build GRCh38 | Loss | FARSA MIR6515 DNASE2 MIR5695 DAND5 KLF1 GCDH GADD45GIP1 NFIX CALR SYCE2 RAD23A |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |