Assay Details
Target Gene Details
Entrez Gene ID: | 79085 |
Gene Name: | solute carrier family 25 member 23 |
Gene Aliases: |
APC2, MCSC2, SCaMC-3 |
Location: |
Chr.19:6436081-6460221 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833727 | Chr.19:6400101 - 6568241 on Build GRCh38 | Loss | KHSRP DENND1C TUBB4A MIR3940 CRB3 SLC25A41 LOC390877 SLC25A23 TNFSF9 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |