Assay Details
Target Gene Details
Entrez Gene ID: | 102724360 |
Gene Name: | uncharacterized LOC102724360 |
Gene Aliases: |
- |
Location: |
Chr.19:18940254-18946831 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC102724360 | NR_134912.1 | |||
NR_134913.1 | ||||
CF127232.1 | ||||
DA366162.1 | ||||
HY133933.1 | ||||
HY154431.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv470132 | Chr.19:18853633 - 18943911 on Build GRCh38 | Loss | GDF1 CERS1 UPF1 DDX49 HOMER3 LOC102724360 COPE |
nsv833778 | Chr.19:18901860 - 19090678 on Build GRCh38 | Loss | SUGP2 SLC25A42 DDX49 HOMER3 LOC102724360 ARMC6 COPE |
nsv1151217 | Chr.19:18627369 - 19284914 on Build GRCh38 | Deletion | HAPLN4 COMP SUGP2 TM6SF2 CERS1 RFXANK UPF1 DDX49 NCAN BORCS8-MEF2B KLHL26 HOMER3 LOC102724360 ARMC6 CRTC1 COPE GDF1 TMEM161A NR2C2AP MEF2B SLC25A42 BORCS8 SUGP1 |