Assay Details
Target Gene Details
Entrez Gene ID: | 1087 |
Gene Name: | carcinoembryonic antigen related cell adhesion molecule 7 |
Gene Aliases: |
CGM2 |
Location: |
Chr.19:41673307-41688277 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CEACAM7 | NM_001291485.1 | NP_001278414.1 | ||
NM_006890.4 | NP_008821.2 | |||
AF006622.1 | AAB62924.1 | |||
AF006623.1 | AAB62925.1 | |||
AK292213.1 | ||||
AK296634.1 | ||||
AK311285.1 | ||||
AK313108.1 | ||||
BC121132.2 | ||||
DC345133.1 | ||||
L31792.1 | AAA66186.1 | |||
X98311.1 | CAA66955.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1061309 | Chr.19:41683785 - 41724525 on Build GRCh38 | Gain | CEACAM5 CEACAM7 |
esv29931 | Chr.19:41682561 - 41687778 on Build GRCh38 | Gain | CEACAM7 |
esv2718577 | Chr.19:41576456 - 41709018 on Build GRCh38 | Deletion | CEACAM5 LOC105372404 CEACAM4 CEACAM7 CEACAM21 |
nsv442464 | Chr.19:41683409 - 41687366 on Build GRCh38 | Gain | CEACAM7 |
esv3644391 | Chr.19:41682404 - 41687675 on Build GRCh38 | Gain | CEACAM7 |
nsv522854 | Chr.19:41613927 - 42426852 on Build GRCh38 | Gain | PRR19 CEACAM5 ZNF526 RPS19 MIR8077 MIR4323 CEACAM3 POU2F2 LYPD4 LOC101930071 CEACAM7 LIPE LOC390937 GSK3A DEDD2 PAFAH1B3 MEGF8 MIR6797 LOC100505585 CIC ERF CD79A LIPE-AS1 ZNF574 CEACAM6 ATP1A3 CEACAM4 CNFN ARHGEF1 TMEM145 GRIK5 LOC100505622 DMRTC2 RABAC1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |