Assay Details
Target Gene Details
Entrez Gene ID: | 2821 |
Gene Name: | glucose-6-phosphate isomerase |
Gene Aliases: |
AMF, GNPI, NLK, PGI, PHI, SA-36, SA36 |
Location: |
Chr.19:34359592-34402413 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GPI | NM_000175.3 | NP_000166.2 | ||
NM_001184722.1 | NP_001171651.1 | |||
NM_001289789.1 | NP_001276718.1 | |||
NM_001289790.1 | NP_001276719.1 | |||
XM_005258764.2 | XP_005258821.1 | |||
XM_006723148.2 | XP_006723211.1 | |||
XM_011526754.2 | XP_011525056.1 | |||
AK025421.1 | ||||
AK129884.1 | ||||
AK130102.1 | ||||
AK293446.1 | ||||
AK294396.1 | ||||
AK301103.1 | ||||
BC004982.1 | AAH04982.1 | |||
K03515.1 | AAA36368.1 | |||
X16539.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3644206 | Chr.19:34296741 - 34419464 on Build GRCh38 | Loss |
![]() ![]() ![]() |
nsv509738 | Chr.19:34351799 - 34412408 on Build GRCh38 | Insertion |
![]() ![]() ![]() |
nsv953279 | Chr.19:34367296 - 34396295 on Build GRCh38 | Deletion |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)