Assay Details
Target Gene Details
Entrez Gene ID: | 23770 |
Gene Name: | FK506 binding protein 8 |
Gene Aliases: |
FKBP38, FKBPr38 |
Location: |
Chr.19:18531752-18544077 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FKBP8 | NM_001308373.1 | NP_001295302.1 | ||
NM_012181.4 | NP_036313.3 | |||
XM_011527863.1 | XP_011526165.1 | |||
XM_011527864.1 | XP_011526166.1 | |||
AK300571.1 | ||||
AK313363.1 | ||||
AY225339.1 | AAO39020.1 | |||
AY278607.1 | AAQ84561.1 | |||
BC009966.1 | AAH09966.1 | |||
BX538124.1 | CAD98028.1 | |||
BX647405.1 | ||||
BX647720.1 | ||||
GQ372970.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833776 | Chr.19:18527218 - 18671454 on Build GRCh38 | Loss | CRLF1 FKBP8 KLHL26 TMEM59L UBA52 C19orf60 KXD1 |
nsv1061733 | Chr.19:18464130 - 18568345 on Build GRCh38 | Gain | FKBP8 UBA52 ELL KXD1 |
nsv1055794 | Chr.19:18500735 - 18566123 on Build GRCh38 | Gain | FKBP8 UBA52 ELL KXD1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |