Assay Details
Target Gene Details
Entrez Gene ID: | 1048 |
Gene Name: | carcinoembryonic antigen related cell adhesion molecule 5 |
Gene Aliases: |
CD66e, CEA |
Location: |
Chr.19:41708585-41730433 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CEACAM5 | NM_001291484.2 | NP_001278413.1 | ||
NM_001308398.1 | NP_001295327.1 | |||
NM_004363.5 | NP_004354.3 | |||
XM_005258413.3 | XP_005258470.2 | |||
XM_011526322.1 | XP_011524624.1 | |||
XM_017026145.1 | XP_016881634.1 | |||
XM_017026146.1 | XP_016881635.1 | |||
AB852566.1 | ||||
AB852567.1 | ||||
AK223101.1 | BAD96821.1 | |||
AK310844.1 | ||||
BC034671.1 | AAH34671.1 | |||
BP272354.1 | ||||
BP272370.1 | ||||
CR749337.1 | CAH18191.1 | |||
DB006625.1 | ||||
M15042.1 | AAA51963.1 | |||
M29540.1 | AAA51967.1 | |||
X16455.1 | CAA34474.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1061309 | Chr.19:41683785 - 41724525 on Build GRCh38 | Gain | CEACAM7 CEACAM5 |
nsv482220 | Chr.19:41708611 - 41730516 on Build GRCh38 | Gain | CEACAM5 |
nsv522854 | Chr.19:41613927 - 42426852 on Build GRCh38 | Gain | POU2F2 PAFAH1B3 ARHGEF1 GSK3A LOC390937 RPS19 MIR4323 DEDD2 LYPD4 LIPE TMEM145 DMRTC2 ZNF574 ATP1A3 ERF GRIK5 LOC100505622 MIR8077 CNFN MEGF8 MIR6797 CEACAM6 CEACAM3 CD79A LIPE-AS1 LOC101930071 CEACAM4 PRR19 RABAC1 CEACAM7 CEACAM5 LOC100505585 ZNF526 CIC |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |