Assay Details
Target Gene Details
Entrez Gene ID: | 117286 |
Gene Name: | calcium and integrin binding family member 3 |
Gene Aliases: |
KIP3 |
Location: |
Chr.19:16161368-16173525 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CIB3 | NM_001300922.1 | NP_001287851.1 | ||
NM_054113.3 | NP_473454.1 | |||
AB050868.1 | BAB71789.1 | |||
BC069320.1 | AAH69320.2 | |||
BC069428.1 | AAH69428.1 | |||
BC069524.1 | AAH69524.1 | |||
BC112200.1 | AAI12201.1 | |||
BC113591.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833767 | Chr.19:16121391 - 16347760 on Build GRCh38 | Loss | KLF2 RAB8A FAM32A AP1M1 HSH2D CIB3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |