Assay Details
Target Gene Details
Entrez Gene ID: | 4784 |
Gene Name: | nuclear factor I X |
Gene Aliases: |
MRSHSS, NF1A, SOTOS2 |
Location: |
Chr.19:12995512-13098796 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NFIX | NM_002501.3 | NP_002492.2 | ||
XM_005259918.4 | XP_005259975.1 | |||
XM_017026837.1 | XP_016882326.1 | |||
BF115883.1 | ||||
BT019732.1 | AAV38537.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv828444 | Chr.19:13012681 - 13027353 on Build GRCh38 | Loss | NFIX |
nsv1055900 | Chr.19:12979147 - 13034197 on Build GRCh38 | Gain | NFIX |
nsv953977 | Chr.19:12666887 - 13095486 on Build GRCh38 | Deletion | C19orf43 GADD45GIP1 ASNA1 TNPO2 HOOK2 MAST1 DAND5 MIR5684 FARSA MIR6794 RNASEH2A BEST2 KLF1 NFIX LOC105372280 JUNB FBXW9 SNORD135 SNORD41 WDR83OS CALR RTBDN SYCE2 DHPS MIR5695 DNASE2 PRDX2 MIR6515 GCDH WDR83 RAD23A |
nsv2422 | Chr.19:13000362 - 13032711 on Build GRCh38 | Insertion | NFIX |
nsv833754 | Chr.19:12876208 - 13028186 on Build GRCh38 | Loss | CALR DAND5 FARSA SYCE2 DNASE2 MIR6515 KLF1 GADD45GIP1 NFIX GCDH MIR5695 RAD23A |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |