Assay Details
Target Gene Details
Entrez Gene ID: | 10780 |
Gene Name: | zinc finger protein 234 |
Gene Aliases: |
HZF4, ZNF269 |
Location: |
Chr.19:44141530-44160309 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ZNF234 | NM_001144824.1 | NP_001138296.1 | ||
NM_006630.2 | NP_006621.1 | |||
XM_006722974.3 | XP_006723037.1 | |||
XM_017026149.1 | XP_016881638.1 | |||
XM_017026150.1 | XP_016881639.1 | |||
XM_017026151.1 | XP_016881640.1 | |||
XM_017026152.1 | XP_016881641.1 | |||
XM_017026153.1 | XP_016881642.1 | |||
AF228417.1 | AAF34785.1 | |||
AK289843.1 | ||||
AK314064.1 | ||||
BC007303.1 | AAH07303.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833840 | Chr.19:44016468 - 44211607 on Build GRCh38 | Gain | ZNF227 ZNF224 ZNF226 ZNF225 ZNF284 ZNF234 ZNF223 LOC100379224 ZNF222 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |