Assay Details
Target Gene Details
Entrez Gene ID: | 4059 |
Gene Name: | basal cell adhesion molecule (Lutheran blood group) |
Gene Aliases: |
AU, CD239, LU, MSK19 |
Location: |
Chr.19:44809059-44821421 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BCAM | NM_001013257.2 | NP_001013275.1 | ||
NM_005581.4 | NP_005572.2 | |||
BC050450.1 | AAH50450.1 | |||
BP352029.1 | ||||
BX387680.2 | ||||
HE572749.1 | ||||
X80026.1 | CAA56327.1 | |||
X83425.1 | CAA58449.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv579862 | Chr.19:44807062 - 44813550 on Build GRCh38 | Gain | BCAM |
nsv579861 | Chr.19:44806715 - 44811138 on Build GRCh38 | Gain | BCAM |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |