Assay Details
Target Gene Details
Entrez Gene ID: | 729767 |
Gene Name: | carcinoembryonic antigen related cell adhesion molecule 18 |
Gene Aliases: |
- |
Location: |
Chr.19:51478543-51491261 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CEACAM18 | NM_001278392.1 | NP_001265321.1 | ||
XM_011527258.2 | XP_011525560.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3644671 | Chr.19:51483799 - 51486019 on Build GRCh38 | Loss | CEACAM18 |