Assay Details
Target Gene Details
Entrez Gene ID: | 94274 |
Gene Name: | protein phosphatase 1 regulatory inhibitor subunit 14A |
Gene Aliases: |
CPI-17, CPI17, PPP1INL |
Location: |
Chr.19:38251237-38256591 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PPP1R14A | NM_001243947.1 | NP_001230876.1 | ||
NM_033256.2 | NP_150281.1 | |||
AB056508.1 | BAB62502.1 | |||
AB056509.1 | BAB62503.1 | |||
AF123763.1 | AAP97242.1 | |||
AY050670.1 | AAL25828.1 | |||
BC021089.1 | AAH21089.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833821 | Chr.19:38139297 - 38266152 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)