Assay Details
Target Gene Details
Entrez Gene ID: | 2901 |
Gene Name: | glutamate ionotropic receptor kainate type subunit 5 |
Gene Aliases: |
EAA2, GRIK2, GluK5, KA2 |
Location: |
Chr.19:41998316-42071804 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GRIK5 | NM_001301030.1 | NP_001287959.1 | ||
NM_002088.4 | NP_002079.3 | |||
XM_005258821.3 | XP_005258878.1 | |||
XM_011526862.2 | XP_011525164.1 | |||
XM_011526863.2 | XP_011525165.1 | |||
XM_011526865.1 | XP_011525167.1 | |||
XM_011526867.1 | XP_011525169.1 | |||
XM_011526868.1 | XP_011525170.1 | |||
XM_011526869.2 | XP_011525171.1 | |||
XM_011526871.2 | XP_011525173.1 | |||
XM_017026713.1 | XP_016882202.1 | |||
AJ249209.1 | CAC80547.1 | |||
S40369.1 | AAB22591.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv520239 | Chr.19:41853387 - 42426852 on Build GRCh38 | Loss | CNFN LIPE-AS1 LOC390937 CIC DEDD2 POU2F2 GRIK5 LIPE CD79A LOC101930071 MIR6797 LOC100505585 MEGF8 MIR8077 GSK3A ZNF526 ERF ARHGEF1 ZNF574 RPS19 TMEM145 RABAC1 MIR4323 PRR19 LOC100505622 PAFAH1B3 ATP1A3 |
nsv522854 | Chr.19:41613927 - 42426852 on Build GRCh38 | Gain | CEACAM3 CNFN LIPE-AS1 LOC390937 CIC DEDD2 POU2F2 DMRTC2 CEACAM6 GRIK5 LIPE CD79A LOC101930071 MIR6797 CEACAM4 LYPD4 CEACAM5 LOC100505585 MEGF8 MIR8077 CEACAM7 GSK3A ZNF526 ERF ARHGEF1 ZNF574 RPS19 TMEM145 RABAC1 MIR4323 PRR19 LOC100505622 PAFAH1B3 ATP1A3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |