Assay Details
Target Gene Details
Entrez Gene ID: | 58513 |
Gene Name: | epidermal growth factor receptor pathway substrate 15 like 1 |
Gene Aliases: |
EPS15R |
Location: |
Chr.19:16355244-16472012 on Build GRCh38 |
Assay Gene Location: | Within Intron 24 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EPS15L1 | NM_001258374.1 | NP_001245303.1 | ||
NM_001258375.1 | NP_001245304.1 | |||
NM_021235.2 | NP_067058.1 | |||
NR_047665.1 | ||||
NR_047666.1 | ||||
XM_017027086.1 | XP_016882575.1 | |||
XM_017027087.1 | XP_016882576.1 | |||
XM_017027088.1 | XP_016882577.1 | |||
XM_017027089.1 | XP_016882578.1 | |||
XM_017027090.1 | XP_016882579.1 | |||
XM_017027091.1 | XP_016882580.1 | |||
AB015346.1 | BAA88118.1 | |||
AF110265.1 | AAF21930.1 | |||
AK024166.1 | ||||
AK025770.1 | ||||
AK291359.1 | ||||
AK296473.1 | ||||
AK297428.1 | ||||
AK299001.1 | ||||
AL110270.1 | CAH10724.1 | |||
BC131590.1 | ||||
BU075732.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)