Assay Details
Target Gene Details
Entrez Gene ID: | 85300 |
Gene Name: | ATCAY, caytaxin |
Gene Aliases: |
BNIP-H, CLAC |
Location: |
Chr.19:3880620-3928082 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ATCAY | NM_033064.4 | NP_149053.1 | ||
AB058775.1 | ||||
AK027889.1 | ||||
AK092309.1 | BAC03859.1 | |||
AK125457.1 | ||||
AK299522.1 | ||||
AY220297.1 | AAO63019.1 | |||
BC026217.1 | AAH26217.1 | |||
BX505461.1 | ||||
DA151785.1 | ||||
DA778097.1 | ||||
KU178814.1 | ||||
KU178815.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833715 | Chr.19:3786758 - 3977144 on Build GRCh38 | Loss | ZFR2 EEF2 MIR637 ATCAY NMRK2 MATK DAPK3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |