Assay Details
Target Gene Details
Entrez Gene ID: | 8725 |
Gene Name: | URI1, prefoldin like chaperone |
Gene Aliases: |
C19orf2, NNX3, PPP1R19, RMP, URI |
Location: |
Chr.19:29923644-30016612 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
URI1 | NM_001252641.1 | NP_001239570.1 | ||
XM_005259362.2 | XP_005259419.1 | |||
XM_011527435.1 | XP_011525737.1 | |||
BC026184.2 | AAH26184.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1063678 | Chr.19:29937920 - 29987524 on Build GRCh38 | Loss | URI1 |
nsv828517 | Chr.19:29781143 - 29950591 on Build GRCh38 | Gain | URI1 CCNE1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |