Assay Details
Target Gene Details
Entrez Gene ID: | 3609 |
Gene Name: | interleukin enhancer binding factor 3 |
Gene Aliases: |
CBTF, DRBF, DRBP76, MMP4, MPHOSPH4, MPP4, NF-AT-90, NF110, NF110b, NF90, NF90a, NF90b, NFAR, NFAR-1, NFAR2, TCP110, TCP80 |
Location: |
Chr.19:10654261-10692419 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1160584 | Chr.19:10618877 - 10892186 on Build GRCh38 | Deletion | MIR4748 SLC44A2 DNM2 MIR199A1 TMED1 C19orf38 ILF3-AS1 QTRT1 ILF3 MIR638 CARM1 MIR6793 |
nsv833745 | Chr.19:10539132 - 10727920 on Build GRCh38 | Loss | MIR1238 SLC44A2 DNM2 KRI1 ILF3-AS1 ATG4D QTRT1 ILF3 MIR638 AP1M2 CDKN2D |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |