Assay Details
Target Gene Details
Entrez Gene ID: | 388551 |
Gene Name: | carcinoembryonic antigen related cell adhesion molecule 16 |
Gene Aliases: |
CEAL2, DFNA4B |
Location: |
Chr.19:44699151-44710714 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CEACAM16 | NM_001039213.3 | NP_001034302.2 | ||
XM_017026795.1 | XP_016882284.1 | |||
EU021223.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833841 | Chr.19:44705217 - 44804786 on Build GRCh38 | Loss | BCL3 CBLC MIR8085 CEACAM16 |
nsv1062812 | Chr.19:44538275 - 44711525 on Build GRCh38 | Gain | CEACAM19 MIR4531 CEACAM22P IGSF23 PVR CEACAM16 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |