Assay Details
Target Gene Details
Entrez Gene ID: | 9244 |
Gene Name: | cytokine receptor like factor 1 |
Gene Aliases: |
CISS, CISS1, CLF, CLF-1, NR6, zcytor5 |
Location: |
Chr.19:18593225-18606850 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CRLF1 | NM_004750.4 | NP_004741.1 | ||
XM_011528422.2 | XP_011526724.1 | |||
XM_011528423.2 | XP_011526725.1 | |||
XM_011528424.2 | XP_011526726.1 | |||
AF059293.1 | AAC28335.1 | |||
AF073515.1 | AAD39681.1 | |||
AF178684.1 | AAD54385.1 | |||
AY358291.1 | AAQ88658.1 | |||
BC044634.1 | AAH44634.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833776 | Chr.19:18527218 - 18671454 on Build GRCh38 | Loss | UBA52 KLHL26 C19orf60 FKBP8 TMEM59L CRLF1 KXD1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |