Assay Details
Target Gene Details
Entrez Gene ID: | 105372412 |
Gene Name: | phospholipase A2 inhibitor and Ly6/PLAUR domain-containing protein-like |
Gene Aliases: |
- |
Location: |
Chr.19:43687557-43700540 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC105372412 | XM_017027562.1 | XP_016883051.1 | ||
XM_017027563.1 | XP_016883052.1 | |||
XM_017027564.1 | XP_016883053.1 | |||
XM_017027565.1 | XP_016883054.1 | |||
XM_017027566.1 | XP_016883055.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3644474 | Chr.19:43687730 - 43702516 on Build GRCh38 | Gain | LOC105372412 |
nsv833839 | Chr.19:43595661 - 43768147 on Build GRCh38 | Loss | ZNF428 ZNF576 LOC105372412 IRGC PLAUR KCNN4 SMG9 SRRM5 CADM4 IRGQ |
esv3893220 | Chr.19:43658802 - 43706225 on Build GRCh38 | Gain | LOC105372412 PLAUR |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |