Assay Details
Target Gene Details
Entrez Gene ID: | 3718 |
Gene Name: | Janus kinase 3 |
Gene Aliases: |
JAK-3, JAK3_HUMAN, JAKL, L-JAK, LJAK |
Location: |
Chr.19:17824782-17848071 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
JAK3 | NM_000215.3 | NP_000206.2 | ||
XM_005259896.3 | XP_005259953.1 | |||
XM_011527990.1 | XP_011526292.1 | |||
XM_011527991.2 | XP_011526293.1 | |||
AK296383.1 | ||||
AK304392.1 | ||||
BC028068.1 | AAH28068.1 | |||
DA405504.1 | ||||
U31601.1 | AAC50226.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |