Assay Details
Target Gene Details
Entrez Gene ID: | 8170 |
Gene Name: | solute carrier family 14 member 2 |
Gene Aliases: |
HUT2, UT-A2, UT2, UTA, UTR, hUT-A6 |
Location: |
Chr.18:45196357-45683686 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC14A2 | NM_001242692.1 | NP_001229621.1 | ||
XM_017026015.1 | XP_016881504.1 | |||
XM_017026016.1 | XP_016881505.1 | |||
AK122698.1 | ||||
AK292422.1 | ||||
DB045789.1 |
Target Gene Details
Entrez Gene ID: | 101927980 |
Gene Name: | SLC14A2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.18:45438183-45507036 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC14A2-AS1 | NR_110899.1 | |||
AK126075.1 | ||||
BX109976.1 | ||||
HY036726.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833638 | Chr.18:45304064 - 45494968 on Build GRCh38 | Gain | SLC14A2-AS1 SLC14A2 |
nsv525268 | Chr.18:45168400 - 45894830 on Build GRCh38 | Gain | EPG5 SIGLEC15 SLC14A2-AS1 SLC14A2 SLC14A1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |