Assay Details
Target Gene Details
Entrez Gene ID: | 54331 |
Gene Name: | G protein subunit gamma 2 |
Gene Aliases: |
- |
Location: |
Chr.14:51847131-51969800 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GNG2 | NM_001243773.1 | NP_001230702.1 | ||
NM_001243774.1 | NP_001230703.1 | |||
NM_053064.4 | NP_444292.1 | |||
XM_006720173.2 | XP_006720236.1 | |||
XM_011536846.2 | XP_011535148.1 | |||
XM_017021376.1 | XP_016876865.1 | |||
XM_017021377.1 | XP_016876866.1 | |||
AK001024.1 | ||||
AK092371.1 | ||||
AK307171.1 | ||||
AK311334.1 | ||||
AL832878.1 | CAI46198.1 | |||
BC020774.1 | AAH20774.1 | |||
BE245271.1 | ||||
DA226565.1 | ||||
DA322879.1 | ||||
DA785895.1 | ||||
DC318652.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv952840 | Chr.14:51846583 - 51949982 on Build GRCh38 | Duplication |
![]() ![]() |
nsv564842 | Chr.14:51803013 - 51867871 on Build GRCh38 | Loss |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)