Assay Details
Target Gene Details
Entrez Gene ID: | 2287 |
Gene Name: | FK506 binding protein 3 |
Gene Aliases: |
FKBP-25, FKBP-3, FKBP25, PPIase |
Location: |
Chr.14:45115599-45134529 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FKBP3 | NM_002013.3 | NP_002004.1 | ||
XM_011536565.2 | XP_011534867.1 | |||
XM_017021092.1 | XP_016876581.1 | |||
XM_017021093.1 | XP_016876582.1 | |||
AK222993.1 | BAD96713.1 | |||
AK311915.1 | ||||
BC016288.1 | AAH16288.1 | |||
BC020809.1 | AAH20809.1 | |||
BI598048.1 | ||||
BT006904.1 | AAP35550.1 | |||
M96256.1 | AAA58471.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832785 | Chr.14:45051467 - 45241538 on Build GRCh38 | Loss | LOC105370474 SNORD127 FANCM PRPF39 MIS18BP1 FAM179B FKBP3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |