Assay Details
Target Gene Details
Entrez Gene ID: | 64430 |
Gene Name: | pecanex homolog 4 (Drosophila) |
Gene Aliases: |
C14orf135, FBP2, PCNXL4 |
Location: |
Chr.14:60091276-60142976 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PCNX4 | NM_022495.5 | NP_071940.4 | ||
XM_005268009.2 | XP_005268066.1 | |||
AK095489.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1044390 | Chr.14:60109151 - 60168783 on Build GRCh38 | Loss | PCNX4 DHRS7 |
esv2759993 | Chr.14:59940776 - 60116323 on Build GRCh38 | Loss | PCNX4 LRRC9 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |