Assay Details
Target Gene Details
Entrez Gene ID: | 3831 |
Gene Name: | kinesin light chain 1 |
Gene Aliases: |
KLC, KNS2, KNS2A |
Location: |
Chr.14:103629188-103701551 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KLC1 | NM_001130107.1 | NP_001123579.1 | ||
NM_005552.4 | NP_005543.2 | |||
NM_182923.3 | NP_891553.2 | |||
AF527736.1 | AAP85635.1 | |||
AK294508.1 | ||||
AY180164.2 | AAO62549.1 | |||
BC008881.2 | AAH08881.1 | |||
BP274336.1 | ||||
DA501532.1 | ||||
L04733.1 | AAA16576.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3635603 | Chr.14:103624778 - 103714856 on Build GRCh38 | Gain | XRCC3 LOC107984648 KLC1 |
nsv832883 | Chr.14:103461350 - 103654616 on Build GRCh38 | Loss | APOPT1 BAG5 TRMT61A KLC1 CKB MARK3 |
nsv566023 | Chr.14:103595309 - 103731914 on Build GRCh38 | Loss | XRCC3 ZFYVE21 LOC107984648 KLC1 |
nsv1054574 | Chr.14:103210111 - 105365114 on Build GRCh38 | Loss | BRF1 LINC00638 LINC00637 BTBD6 TDRD9 LOC388022 LOC102723354 LOC105370708 MIR203A CKB LOC105370691 CDCA4 ZBTB42 XRCC3 APOPT1 MIR6765 ZFYVE21 MIR203B TRMT61A C14orf79 NUDT14 SNORA28 PACS2 ADSSL1 BAG5 AKT1 LOC101929634 PLD4 LOC107984648 LOC105370687 RD3L KLC1 CEP170B SIVA1 EIF5 MIR4710 C14orf180 MARK3 C14orf2 INF2 GPR132 AHNAK2 TMEM179 JAG2 LOC105378183 PPP1R13B KIF26A ASPG |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |