Assay Details
Target Gene Details
Entrez Gene ID: | 145482 |
Gene Name: | prostaglandin reductase 2 |
Gene Aliases: |
HEL-S-298, PGR2, ZADH1 |
Location: |
Chr.14:73851831-73885465 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PTGR2 | NM_001146154.1 | NP_001139626.1 | ||
NM_001146155.1 | NP_001139627.1 | |||
NM_152444.2 | NP_689657.1 | |||
AK096410.1 | BAC04781.1 | |||
AY346133.1 | AAR05101.1 | |||
AY424308.1 | AAR82927.1 | |||
BC059364.1 | AAH59364.1 | |||
BP357520.1 | ||||
BX641118.1 | CAE46055.1 | |||
DR158023.1 | ||||
FJ224334.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1053281 | Chr.14:73518500 - 74050155 on Build GRCh38 | Gain | ZNF410 ACOT2 PNMA1 MIR4505 DNAL1 ENTPD5 ACOT1 ACOT4 LOC100506476 FAM161B PTGR2 LOC100506498 COQ6 ELMSAN1 BBOF1 ACOT6 HEATR4 |
nsv1035460 | Chr.14:72854572 - 73897173 on Build GRCh38 | Gain | ZNF410 ACOT2 PNMA1 LOC105370561 RBM25 MIR4505 DNAL1 ACOT1 ACOT4 LOC100506476 PTGR2 PSEN1 NUMB LOC100506498 LOC101928143 ELMSAN1 C14orf169 DPF3 LOC101928123 ACOT6 PAPLN HEATR4 DCAF4 ZFYVE1 |
nsv976844 | Chr.14:73850066 - 73884634 on Build GRCh38 | Duplication | PTGR2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |