Assay Details
Target Gene Details
Entrez Gene ID: | 63874 |
Gene Name: | abhydrolase domain containing 4 |
Gene Aliases: |
ABH4 |
Location: |
Chr.14:22598238-22612864 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ABHD4 | NM_022060.2 | NP_071343.2 | ||
XM_005267986.4 | XP_005268043.1 | |||
AK022878.1 | BAB14289.1 | |||
AK024790.1 | ||||
AK293198.1 | ||||
AK298127.1 | ||||
BC024779.1 | AAH24779.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832746 | Chr.14:22506563 - 22681586 on Build GRCh38 | Gain | LOC105370399 ABHD4 TRAC TRAJ17 DAD1 |
nsv563998 | Chr.14:22414183 - 22879743 on Build GRCh38 | Gain | LRP10 MRPL52 TRAJ60 OXA1L MMP14 LOC105370399 ABHD4 TRDV2 TRAC TRAJ17 DAD1 SLC7A7 TRDV3 |
nsv832748 | Chr.14:22515242 - 22630030 on Build GRCh38 | Gain | LOC105370399 ABHD4 TRAC TRAJ17 DAD1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |