Assay Details
Target Gene Details
Entrez Gene ID: | 4149 |
Gene Name: | MYC associated factor X |
Gene Aliases: |
bHLHd4 |
Location: |
Chr.14:65006101-65102695 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MAX | NM_001271069.1 | NP_001257998.1 | ||
NM_197957.3 | NP_932061.1 | |||
AI652319.1 | ||||
AI655674.1 | ||||
AI917842.1 | ||||
DB482510.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832816 | Chr.14:65006526 - 65177927 on Build GRCh38 | Gain | CHURC1-FNTB FNTB LOC100506321 MIR4706 MAX |
esv2748779 | Chr.14:63663324 - 66595809 on Build GRCh38 | Deletion | CHURC1-FNTB GPX2 ESR2 LINC00238 MIR548H1 MIR4706 ZBTB25 PLEKHG3 AKAP5 PPP1R36 MIR7855 ZBTB1 FUT8-AS1 HSPA2 LOC100128233 FNTB CHURC1 SPTB SYNE2 MAX SGPP1 MTHFD1 LOC100506321 TEX21P GPHN MIR625 FUT8 MIR4708 LOC102723809 RAB15 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |