Assay Details
Target Gene Details
Entrez Gene ID: | 145482 |
Gene Name: | prostaglandin reductase 2 |
Gene Aliases: |
HEL-S-298, PGR2, ZADH1 |
Location: |
Chr.14:73851831-73885465 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PTGR2 | NM_001146154.1 | NP_001139626.1 | ||
NM_001146155.1 | NP_001139627.1 | |||
NM_152444.2 | NP_689657.1 | |||
AK096410.1 | BAC04781.1 | |||
AY346133.1 | AAR05101.1 | |||
AY424308.1 | AAR82927.1 | |||
BC059364.1 | AAH59364.1 | |||
BP357520.1 | ||||
BX641118.1 | CAE46055.1 | |||
DR158023.1 | ||||
FJ224334.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1053281 | Chr.14:73518500 - 74050155 on Build GRCh38 | Gain | ZNF410 DNAL1 ELMSAN1 ACOT2 FAM161B LOC100506476 HEATR4 MIR4505 PTGR2 PNMA1 LOC100506498 ENTPD5 ACOT1 COQ6 ACOT6 ACOT4 BBOF1 |
nsv1035460 | Chr.14:72854572 - 73897173 on Build GRCh38 | Gain | ZNF410 PAPLN DNAL1 ELMSAN1 RBM25 ZFYVE1 C14orf169 ACOT2 LOC105370561 LOC100506476 HEATR4 MIR4505 NUMB LOC101928123 PTGR2 PNMA1 DPF3 DCAF4 LOC100506498 ACOT1 PSEN1 ACOT6 ACOT4 LOC101928143 |
nsv976844 | Chr.14:73850066 - 73884634 on Build GRCh38 | Duplication | PTGR2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |